Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Birch pollen Bet V1, Rabbit anti-Plant, Clone: BV16, Novus Biologicals™
Rabbit Monoclonal Antibody
$265.00 - $566.50
Specifications
Antigen | Birch pollen Bet V1 |
---|---|
Clone | BV16 |
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA |
Applications | Western Blot, ELISA |
Description
Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.Specifications
Birch pollen Bet V1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Plant | |
Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
IgG κ | |
Protein A purified | |
This antibody binds to Birch pollen Bet V1. |
BV16 | |
Western Blot, ELISA | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
Birch pollen Bet V1-A, a major allergen of the European white birch. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title