Alfa Aesar™ Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

Manufacturer: Alfa Aesar™ 3941499

Quantity 5g

 View more versions of this product

Catalog No. AA3941499

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35138-23-9
    Formula Weight 495.4
    MDL Number MFCD00079664
    Chemical Name or Material Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

    Specifications

    Formula Weight 495.4
    Quantity 5g
    Solubility Information Soluble in chloroform, methylene chloride
    Physical Form Powder
    MDL Number MFCD00079664
    CAS 35138-23-9
    Chemical Name or Material Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit