Learn More
Alfa Aesar™ Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Manufacturer:  Alfa Aesar™ 39414ME
Quantity 500mg
 View more versions of this product
Catalog No. AA39414ME
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
35138-23-9
495.4
MFCD00079664
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Specifications
35138-23-9
495.4
500mg
Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Powder
MFCD00079664
Soluble in chloroform, methylene chloride