Alfa Aesar™ Bis(1,5-cyclooctadiene)nickel(0)

Manufacturer: Alfa Aesar™ 3923402

Quantity 0.5g

 View more versions of this product

Catalog No. AA3923402

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1295-35-8
    Formula Weight 275.06
    MDL Number MFCD00058902
    UN Number UN1325
    Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)

    Specifications

    Formula Weight 275.06
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number QR6135000
    Physical Form Crystalline
    MDL Number MFCD00058902
    UN Number UN1325
    Color Yellow
    CAS 1295-35-8
    Sensitivity Air & Light Sensitive
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Melting Point 60°C (dec.)
    Quantity 0.5g
    Health Hazard 2 GHS H Statement
    H228-H350-H317
    Flammable solid.
    May cause cancer.
    May cause an allergic skin reaction.
    Odor Odorless
    Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)

    GHS H Statement
    H228-H350-H317
    Flammable solid.
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P363-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit