Learn More
Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals
Description
Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and is used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. Also, it is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 60°C (decomposition) |
Color | Yellow |
Odor | Odorless |
Quantity | 2 g |
UN Number | UN1325 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in benzene,toluene,terahydrofuran,ether,dimethyl formamide,hexamethylphosphoramide,N-methylpyrrolidinone. Insoluble in water. |
IUPAC Name | bis((1Z,5Z)-cycloocta-1,5-diene) nickel |
Formula Weight | 275.06 |
Physical Form | Crystalline |
Show More |
Safety and Handling
Hazard Category | H228-H317-H350 |
Hazard Statement | GHS H Statement H228-H350-H317 Flammable solid. May cause cancer. May cause an allergic skin reaction. |
Precautionary Statement | P201-P202-P210-P240-P241-P261-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 215-072-0 |
RTECSNumber | QR6135000 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.