Alfa Aesar™ Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate

Manufacturer: Alfa Aesar™ 45523MD

Quantity 250mg

 View more versions of this product

Catalog No. AA45523MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 130296-28-5
    Molecular Formula C16H24F6RhSb
    Formula Weight 555.01
    MDL Number MFCD11113031
    UN Number UN3077
    Chemical Name or Material Bis (1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate

    Specifications

    Physical Form Crystalline Powder
    Formula Weight 555.01
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 130296-28-5
    Quantity 250mg
    MDL Number MFCD11113031
    UN Number UN3077
    Color Brown
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Molecular Formula C16H24F6RhSb
    Health Hazard 1 Warning
    Chemical Name or Material Bis (1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit