Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
35138-22-8
C16H24Rh·BF4
406.08
MFCD00075045
LYXHWHHENVLYCN-QMDOQEJBSA-N
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate,rhodium i tetrafluoroborate 1,5-cyclooctadiene complex,1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate,bis 1,5-cyclooctadiene rhodium tetrafluoroborate,rh cod 2 bf4,c16h24bf4rh,c16h24rh.bf4,bis 1,5-cyclooctadien rhodium i tetrafluoroborate
5702662
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1.[Rh]
35138-22-8
C16H24Rh·BF4
406.08
MFCD00075045
LYXHWHHENVLYCN-QMDOQEJBSA-N
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate,rhodium i tetrafluoroborate 1,5-cyclooctadiene complex,1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate,bis 1,5-cyclooctadiene rhodium tetrafluoroborate,rh cod 2 bf4,c16h24bf4rh,c16h24rh.bf4,bis 1,5-cyclooctadien rhodium i tetrafluoroborate
5702662
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1.[Rh]
Melting Point 210.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250mg
Color Red
Linear Formula [Rh(cod)2]BF4
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
Formula Weight 406.08
Physical Form Powder
Chemical Name or Material Bis(1, 5-Cyclooctadiene)rhodium(I) tetrafluoroborate

GHS H Statement
Flammable solid.
May be corrosive to metals.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep only in original container.
Absorb spillage to prevent material damage.

GHS Signal Word: Danger