Alfa Aesar™ Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Manufacturer: Alfa Aesar™ 39289ME

Quantity 500mg

 View more versions of this product

Catalog No. AA39289ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99326-34-8
    Formula Weight 468.34
    MDL Number MFCD00143752
    Synonym Bis(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate
    Chemical Name or Material Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

    Specifications

    Formula Weight 468.34
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Physical Form Crystalline
    Synonym Bis(1,5-cyclooctadiene)rhodium(I)trifluoromethanesulfonate
    Solubility Information Soluble in chloroform
    Health Hazard 1 Warning
    MDL Number MFCD00143752
    Quantity 500mg
    Sensitivity Air Sensitive
    Storage Note 1
    • Packaged under argon
    CAS 99326-34-8
    Chemical Name or Material Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit