Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Click to view available options
Quantity:
500mg
2g
99326-34-8
C17H24F3O3RhS-
468.337
MFCD00143752
VUTUHLLWFPRWMT-QMDOQEJBSA-M
bis 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,bis 1,5-cyclooctadiene rhodium i triflate,rh cod 2 otf,bis 1,5-cyclooctadiene rhodium i trifl&,bis 1,5-cyclooctadiene rhodium triflate,rhodium 1+ ion bis 1,5-cyclooctadiene triflate,bis 1,5-cyclooctadiene rhodium i trifluoromethane,n-tert-butyloxycarbonyl-n'-benzyloxymethyl-l-histidine,bis 1,5-cyclooctadiene-trifluoromethanesulfitorhodium i
5702638
(1Z,5Z)-cycloocta-1,5-diene;rhodium;trifluoromethanesulfonate
C1CC=CCCC=C1.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
99326-34-8
C17H24F3O3RhS-
468.337
MFCD00143752
VUTUHLLWFPRWMT-QMDOQEJBSA-M
bis 1,5-cyclooctadiene rhodium i trifluoromethanesulfonate,bis 1,5-cyclooctadiene rhodium i triflate,rh cod 2 otf,bis 1,5-cyclooctadiene rhodium i trifl&,bis 1,5-cyclooctadiene rhodium triflate,rhodium 1+ ion bis 1,5-cyclooctadiene triflate,bis 1,5-cyclooctadiene rhodium i trifluoromethane,n-tert-butyloxycarbonyl-n'-benzyloxymethyl-l-histidine,bis 1,5-cyclooctadiene-trifluoromethanesulfitorhodium i
5702638
(1Z,5Z)-cycloocta-1,5-diene;rhodium;trifluoromethanesulfonate
C1CC=CCCC=C1.C1CC=CCCC=C1.C(F)(F)(F)S(=O)(=O)[O-].[Rh]
  • Packaged under argon
Quantity 2g
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform
Formula Weight 468.34
Physical Form Crystalline
Chemical Name or Material Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures