Alfa Aesar™ Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%

Manufacturer: Alfa Aesar™ H5556214

Quantity 25g

 View more versions of this product

Catalog No. AAH5556214

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 92466-70-1
    Linear Formula (CF3CH2O)2P(O)H
    Formula Weight 246.05
    MDL Number MFCD00063314
    Synonym Phosphonic acid bis(2,2,2-trifluoroethyl) ester
    Chemical Name or Material Bis(2,2,2-trifluoroethyl) phosphite
    Grade Technical Grade

    Specifications

    Formula Weight 246.05
    Boiling Point 43° to 44°C (2mmHg)
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.3320
    Physical Form Liquid
    CAS 92466-70-1
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Density 1.545g/mL
    MDL Number MFCD00063314
    Health Hazard 1 Warning
    Quantity 25g
    Linear Formula (CF3CH2O)2P(O)H
    Synonym Phosphonic acid bis(2,2,2-trifluoroethyl) ester
    Chemical Name or Material Bis(2,2,2-trifluoroethyl) phosphite
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

    Notes
    Store at room temperature. Avoid strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit