Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate, 98+%

MOCVD of superconductors

Manufacturer: Alfa Aesar™ 1453906

Quantity 5g

 View more versions of this product

Catalog No. AA1453906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17594-47-7
    Formula Weight 503.85 (Anhydrous)
    MDL Number MFCD00058709
    UN Number UN1564
    Synonym Barium-DPM hydrate; Ba(TMHD)2
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate

    Specifications

    Formula Weight 503.85 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    MDL Number MFCD00058709
    UN Number UN1564
    Color White
    Melting Point 172°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Boiling Point 225°C (0.3mmHg)
    Health Hazard 1 Warning
    Sensitivity Hygroscopic
    Synonym Barium-DPM hydrate; Ba(TMHD)2
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    CAS 17594-47-7
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate
    Assay ≥98%

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit