Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II), 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4158918

Quantity 50g

 View more versions of this product

Catalog No. AA4158918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36818-89-0
    Formula Weight 406.57
    MDL Number MFCD00064757
    Synonym Calcium-DPM; Ca(TMHD)2
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)

    Specifications

    Formula Weight 406.57
    Physical Form Powder
    Synonym Calcium-DPM; Ca(TMHD)2
    Boiling Point 205°C (0.1mmHg) (subl.)
    Sensitivity Hygroscopic
    MDL Number MFCD00064757
    CAS 36818-89-0
    Melting Point 202° to 206°C
    Quantity 50g
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)
    Assay 99.99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit