Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis)
Click to view available options
Quantity:
0.5g
2g
10g
Specifications
Melting Point | 143°C |
Quantity | 0.5g |
Solubility Information | Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water. |
Formula Weight | 425.46 |
Physical Form | Powder |
Percent Purity | 99.9% |
Chemical Name or Material | Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341
TSCA : No
Recommended Storage : Ambient temperatures