Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

Manufacturer: Alfa Aesar™ 3939703

Quantity 1g

 View more versions of this product

Catalog No. AA3939703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21319-43-7
    Formula Weight 573.75
    MDL Number MFCD00070468
    UN Number UN3467
    Synonym Lead(II)-DPM; Pb(TMHD)2
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

    Specifications

    Formula Weight 573.75
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 126° to 128°C
    Quantity 1g
    MDL Number MFCD00070468
    UN Number UN3467
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder
    CAS 21319-43-7
    Synonym Lead(II)-DPM; Pb(TMHD)2
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
    Health Hazard 1 Danger
    Boiling Point 134°C (0.1mmHg) (subl.)
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit