Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%

NMR shift reagent, nickel oxide precursor

Manufacturer: Alfa Aesar™ 4047014

Quantity 25g

 View more versions of this product

Catalog No. AA4047014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14481-08-4
    Formula Weight 425.24
    MDL Number MFCD00192348
    Synonym Nickel(II)-DPM; Ni(TMHD)2
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)

    Specifications

    Formula Weight 425.24
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    Physical Form Powder
    CAS 14481-08-4
    Synonym Nickel(II)-DPM; Ni(TMHD)2
    Melting Point 223° to 225°C
    Health Hazard 1 Danger
    MDL Number MFCD00192348
    Boiling Point 90° to 110°C (0.1mmHg) (subl.)
    Health Hazard 2 GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.
    Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)
    Assay 98%

    GHS H Statement
    H350-H317
    May cause cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P281-P363-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit