Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane, 95+%, Acros Organics

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
89597-05-7
C20H32Si
300.56
MFCD00674017
ZGFFKHWCLRFAAX-UHFFFAOYSA-N
dimethylbis 2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl silane,dimethyl-bis 2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl silane,dimethylbis 2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl silane,acmc-20al65,silane, dimethylbis tetramethylcyclopentadienyl-mixed isomers,dimethylbis 2,3,4,5-tetramethylcyclopentadienyl silane,dimethyldi 2,3,4,5-tetramethyl-2,4-cyclopentadienyl silane,bis 2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl dimethylsilane,bis-2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl-dimethylsilane,dimethyl-bis 2,3,4,5-tetramethyl-1-cyclopenta-2,4-dienyl silane
601539
dimethyl-bis(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silane
CC1=C(C(=C(C1[Si](C)(C)C2C(=C(C(=C2C)C)C)C)C)C)C
89597-05-7
C20H32Si
300.56
MFCD00674017
ZGFFKHWCLRFAAX-UHFFFAOYSA-N
dimethylbis 2,3,4,5-tetramethyl-2,4-cyclopentadien-1-yl silane,dimethyl-bis 2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl silane,dimethylbis 2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl silane,acmc-20al65,silane, dimethylbis tetramethylcyclopentadienyl-mixed isomers,dimethylbis 2,3,4,5-tetramethylcyclopentadienyl silane,dimethyldi 2,3,4,5-tetramethyl-2,4-cyclopentadienyl silane,bis 2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl dimethylsilane,bis-2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl-dimethylsilane,dimethyl-bis 2,3,4,5-tetramethyl-1-cyclopenta-2,4-dienyl silane
601539
dimethyl-bis(2,3,4,5-tetramethylcyclopenta-2,4-dien-1-yl)silane
CC1=C(C(=C(C1[Si](C)(C)C2C(=C(C(=C2C)C)C)C)C)C)C
Packaging Glass bottle
Melting Point 65.0°C to 68.0°C
Quantity 500mg
Infrared Spectrum Authentic
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Formula Weight 300.56
Physical Form Powder and Chunks
Percent Purity 95+%
Chemical Name or Material Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
Show More Show Less