Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(2-chloroethyl)methylamine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H5506603

Quantity 1g

 View more versions of this product

Catalog No. AAH5506603

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

55-86-7
(ClCH2CH2)2NCH3·HCl
MFCD00012517
14,5775
Bis(2-chloroethyl)methylamine hydrochloride
C5H11Cl2N·HCl
192.52
UN2923
Mechlorethamine hydrochloride; Chlormethine hydrochloride

Specifications

55-86-7
(ClCH2CH2)2NCH3·HCl
MFCD00012517
108° to 111°C
Mechlorethamine hydrochloride; Chlormethine hydrochloride
Bis(2-chloroethyl)methylamine hydrochloride
C5H11Cl2N·HCl
192.52
UN2923
14,5775
1g
98%

Safety and Handling

GHS H Statement
H300-H334-H340-H350-H314-H318-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : IA2100000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit