Alfa Aesar™ Bis(2-chloroethyl)methylamine hydrochloride, 98%

Manufacturer: Alfa Aesar™ H5506614

Quantity 25g

 View more versions of this product

Catalog No. AAH5506614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 55-86-7
    Molecular Formula C5H11Cl2N·HCl
    Linear Formula (ClCH2CH2)2NCH3·HCl
    Formula Weight 192.52
    MDL Number MFCD00012517
    UN Number UN2923
    Merck Index 14,5775
    Synonym Mechlorethamine hydrochloride; Chlormethine hydrochloride
    Chemical Name or Material Bis(2-chloroethyl)methylamine hydrochloride

    Specifications

    Formula Weight 192.52
    TSCA TSCA
    Linear Formula (ClCH2CH2)2NCH3·HCl
    MDL Number MFCD00012517
    UN Number UN2923
    Melting Point 108° to 111°C
    Merck Index 14,5775
    Synonym Mechlorethamine hydrochloride; Chlormethine hydrochloride
    Quantity 25g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H334-H340-H350-H314-H318-H317
    Fatal if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number IA2100000
    Molecular Formula C5H11Cl2N·HCl
    CAS 55-86-7
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material Bis(2-chloroethyl)methylamine hydrochloride
    Assay 98%

    GHS H Statement
    H300-H334-H340-H350-H314-H318-H317
    Fatal if swallowed.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit