Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-diphenylphosphinophenyl)ether, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
166330-10-5
C36H28OP2
538.56
MFCD00233863
RYXZOQOZERSHHQ-UHFFFAOYSA-N
bis 2-diphenylphosphinophenyl ether,dpephos,oxydi-2,1-phenylene bis diphenylphosphine,oxybis 2,1-phenylene bis diphenylphosphine,bis 2-diphenylphosphino phenyl ether,2,2'-bis diphenylphosphino diphenyl ether,2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
4285986
[2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3OC4=CC=CC=C4P(C5=CC=CC=C5)C6=CC=CC=C6
166330-10-5
C36H28OP2
538.56
MFCD00233863
RYXZOQOZERSHHQ-UHFFFAOYSA-N
bis 2-diphenylphosphinophenyl ether,dpephos,oxydi-2,1-phenylene bis diphenylphosphine,oxybis 2,1-phenylene bis diphenylphosphine,bis 2-diphenylphosphino phenyl ether,2,2'-bis diphenylphosphino diphenyl ether,2-2-diphenylphosphanyl phenoxy phenyl diphenylphosphane
4285986
[2-(2-diphenylphosphanylphenoxy)phenyl]-diphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3OC4=CC=CC=C4P(C5=CC=CC=C5)C6=CC=CC=C6
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 184.0°C to 187.0°C
Color White
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula [(C6H5)2P(C6H4)]2O
Solubility Information Solubility in water: insoluble
Formula Weight 538.56
Physical Form Crystalline Powder or Crystals
Percent Purity 99%
Chemical Name or Material Bis(2-diphenylphosphinophenyl)ether
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning