Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(2-ethylhexyl) phosphate, 95%

Extraction of metals

Manufacturer:  Alfa Aesar™ 17723A3

Quantity 2kg

 View more versions of this product

Catalog No. AA17723A3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

298-07-7
322.43
UN1902
Bis(2-ethylhexyl) phosphate
[CH3(CH2)3CH(C2H5)CH2O]2P(O)(OH)
MFCD00009492
DEHPA; Di(2-ethylhexyl)phosphoric acid

Specifications

298-07-7
95%
[CH3(CH2)3CH(C2H5)CH2O]2P(O)(OH)
MFCD00009492
DEHPA; Di(2-ethylhexyl)phosphoric acid
Colorless
2 to 3
2kg
Bis(2-ethylhexyl) phosphate
Liquid
1.4420
322.43
UN1902
0.965g/mL
−50°C
48°C (12mmHg)
171° (340°F)
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: DIISOOCTYL ACID PHOSPHATE

RTECSNumber : TB7875000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit