Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethylene glycol, 99%, Acros Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
111-46-6
C4H10O3
106.12
MFCD00002882
MTHSVFCYNBDYFN-UHFFFAOYSA-N
diethylene glycol,2,2'-oxydiethanol,diglycol,diethylenglykol,2-hydroxyethyl ether,bis 2-hydroxyethyl ether,ethanol, 2,2'-oxybis,2,2'-oxybisethanol,2-2-hydroxyethoxy ethanol,digol
8117
CHEBI:46807
2-(2-hydroxyethoxy)ethanol
C(COCCO)O
111-46-6
C4H10O3
106.12
MFCD00002882
MTHSVFCYNBDYFN-UHFFFAOYSA-N
diethylene glycol,2,2'-oxydiethanol,diglycol,diethylenglykol,2-hydroxyethyl ether,bis 2-hydroxyethyl ether,ethanol, 2,2'-oxybis,2,2'-oxybisethanol,2-2-hydroxyethoxy ethanol,digol
8117
CHEBI:46807
2-(2-hydroxyethoxy)ethanol
C(COCCO)O
Viscosity 38 mPa.s (20°C)
Melting Point -10.0°C
Water 0.2% max.
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4450 to 1.4480
Flash Point 143°C
Quantity 2.5L
Specific Gravity 1.11
Density 1.1100g/mL
Boiling Point 245.0°C
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (HOCH2CH2)2O
Beilstein 01, 468
Merck Index 15, 3131
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, ether,, ethylene glycol, insoluble in benzene, toluene, carbon, tetrachloride and aliphatic hydrocarbons
Formula Weight 106.12
Physical Form Oily Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Diethylene glycol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning