Alfa Aesar™ Bis(2-methoxyethyl) adipate, 98%

Manufacturer: Alfa Aesar™ A1049722

Quantity 100g

 View more versions of this product

Catalog No. AAA1049722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-00-3
    Molecular Formula C12H22O6
    Linear Formula (CH2)4(CO2CH2CH2OCH3)2
    Formula Weight 262.3
    MDL Number MFCD00041905
    Synonym Adipic acid bis(2-methoxyethyl) ester
    Chemical Name or Material Bis (2-methoxyethyl) adipate

    Specifications

    Formula Weight 262.3
    Molecular Formula C12H22O6
    Linear Formula (CH2)4(CO2CH2CH2OCH3)2
    MDL Number MFCD00041905
    CAS 106-00-3
    TSCA TSCA
    Synonym Adipic acid bis(2-methoxyethyl) ester
    Boiling Point 190°C (11mmHg)
    Quantity 100g
    Chemical Name or Material Bis (2-methoxyethyl) adipate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit