Learn More
[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 368961000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
[Bis(2-methoxyethyl)amino]sulfur trifluoride | |
49.0 | |
Orange-Yellow | |
4°C | |
C6H14F3NO2S | |
100 g | |
15,2899 | |
APOYTRAZFJURPB-UHFFFAOYSA-N | |
2-methoxy-N-(2-methoxyethyl)-N-(trifluoro-$l^{4}-sulfanyl)ethanamine | |
2734690 | |
≥95% (NMR, neat substance) |
202289-38-1, 108-88-3 | |
51.0 | |
1.2000g/mL | |
Plastic bottle | |
(CH3OCHCH2)2NSF3 | |
1.2 | |
bis 2-methoxyethyl aminosulfur trifluoride, deoxo-fluor, 2-methoxy-n-2-methoxyethyl-n-trifluorothio-ethanamine, deoxo-fluor r, bis 2-methoxyethyl aminosulphurtrifluoride, bast, bis 2-methoxyethyl amino sulfur trifluoride, bis 2-methoxyethyl aminosulphur trifluoride | |
COCCN(CCOC)S(F)(F)F | |
221.24 | |
221.24 | |
Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Reacts violently with water.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Store in a dry place. Store in a closed container. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only