Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyethyl) ether, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
111-96-6
C6H14O3
134.18
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane
8150
CHEBI:46784
1-methoxy-2-(2-methoxyethoxy)ethane
COCCOCCOC
111-96-6
C6H14O3
134.18
SBZXBUIDTXKZTM-UHFFFAOYSA-N
diglyme,bis 2-methoxyethyl ether,2-methoxyethyl ether,1-methoxy-2-2-methoxyethoxy ethane,diethylene glycol dimethyl ether,poly-solv,di 2-methoxyethyl ether,dimethyl carbitol,2,5,8-trioxanonane
8150
CHEBI:46784
1-methoxy-2-(2-methoxyethoxy)ethane
COCCOCCOC
Viscosity 2 mPa.s (20°C)
Specific Gravity 0.937
Water 0.025% max.
Refractive Index 1.4070 to 1.4090
Density 0.9370g/mL
Packaging Glass bottle
Melting Point -64.0°C
Color Undesignated
Quantity 250mL
Infrared Spectrum Authentic
Flash Point 51°C
Boiling Point 162.0°C
Assay Percent Range 99%
Linear Formula (CH3OCH2CH2)2O
Beilstein 01,IV,2393
Fieser 01,255
Merck Index 15,3184
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,ether,hydrocarbon solvents
Formula Weight 134.18
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Bis(2-methoxyethyl) ether
Show More Show Less