Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyphenyl)phenylphosphine, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
36802-41-2
C20H19O2P
322.344
MFCD00061478
MVKZAEARORRRPG-UHFFFAOYSA-N
bis 2-methoxyphenyl phenylphosphine,bis 2-methoxyphenyl phenyl phosphine,bis 2-methoxyphenyl phenyl phosphane,bis o-anisyl phenylphosphine,bis 2-methoxyphenyl-phenylphosphane,bis-2-methoxyphenyl phenylphosphine,phosphine,bis 2-methoxyphenyl phenyl,phosphine, bis 2-methoxyphenyl phenyl,bis-2-methoxy-phenyl-phenyl-phosphane
4405408
bis(2-methoxyphenyl)-phenylphosphane
COC1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3OC
36802-41-2
C20H19O2P
322.344
MFCD00061478
MVKZAEARORRRPG-UHFFFAOYSA-N
bis 2-methoxyphenyl phenylphosphine,bis 2-methoxyphenyl phenyl phosphine,bis 2-methoxyphenyl phenyl phosphane,bis o-anisyl phenylphosphine,bis 2-methoxyphenyl-phenylphosphane,bis-2-methoxyphenyl phenylphosphine,phosphine,bis 2-methoxyphenyl phenyl,phosphine, bis 2-methoxyphenyl phenyl,bis-2-methoxy-phenyl-phenyl-phosphane
4405408
bis(2-methoxyphenyl)-phenylphosphane
COC1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3OC
Color White
Melting Point 158°C to 165°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 322.34
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Bis(2-methoxyphenyl)phenylphosphine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.