Alfa Aesar™ Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

Manufacturer: Alfa Aesar™ H2689103

Quantity 1g

 View more versions of this product

Catalog No. AAH2689103

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 12289-94-0
    Molecular Formula C16H26Ru
    Formula Weight 319.45
    MDL Number MFCD00216965
    UN Number UN3395
    Synonym (1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium(II); Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex
    Chemical Name or Material Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

    Specifications

    Formula Weight 319.45
    Melting Point 83° to 86°C
    Quantity 1g
    Molecular Formula C16H26Ru
    MDL Number MFCD00216965
    CAS 12289-94-0
    UN Number UN3395
    Synonym (1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium(II); Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H261
    In contact with water releases flammable gas.
    Health Hazard 3 GHS P Statement
    P231+P232-P280-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    DOT Information Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE
    Chemical Name or Material Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

    Description

    Applications
    Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.

    Solubility
    It is soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

    GHS H Statement
    H261
    In contact with water releases flammable gas.

    −30° to −10°C

    GHS P Statement
    P231+P232-P280-P370+P378c-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, sand, extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit