Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-Nitrophenyl) disulfide, 95%, Acros Organics

Click to view available options
Quantity:
5g
1155-00-6
C12H8N2O4S2
308.326
MFCD00007130
NXCKJENHTITELM-UHFFFAOYSA-N
bis 2-nitrophenyl disulfide,disulfide, bis 2-nitrophenyl,2,2'-dinitrodiphenyl disulfide,o-nitrophenyl disulfide,2-nitrophenyl disulfide,o,o'-dinitrodiphenyl disulfide,disulfide, bis o-nitrophenyl,bis o-nitrophenyl disulfide,1,2-bis 2-nitrophenyl disulfane
70861
1-nitro-2-[(2-nitrophenyl)disulfanyl]benzene
C1=CC=C(C(=C1)[N+](=O)[O-])SSC2=CC=CC=C2[N+](=O)[O-]
1155-00-6
C12H8N2O4S2
308.326
MFCD00007130
NXCKJENHTITELM-UHFFFAOYSA-N
bis 2-nitrophenyl disulfide,disulfide, bis 2-nitrophenyl,2,2'-dinitrodiphenyl disulfide,o-nitrophenyl disulfide,2-nitrophenyl disulfide,o,o'-dinitrodiphenyl disulfide,disulfide, bis o-nitrophenyl,bis o-nitrophenyl disulfide,1,2-bis 2-nitrophenyl disulfane
70861
1-nitro-2-[(2-nitrophenyl)disulfanyl]benzene
C1=CC=C(C(=C1)[N+](=O)[O-])SSC2=CC=CC=C2[N+](=O)[O-]
Quantity 5g
Infrared Spectrum Authentic
Color Undesignated
Melting Point 188°C to 195°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (O2NC6H4)2S2
Beilstein 06, 338
Formula Weight 308.32
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material Bis(2-Nitrophenyl) disulfide, 95%
Show More Show Less