Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(2-thiophenemethyl)amine hydrochloride, 99%

Click to view available options
Quantity:
10g
50g
336624-10-3
C10H12ClNS2
245.783
MFCD01812493
RTJOHEYKSNEPMI-UHFFFAOYSA-N
bis 2-thiophenemethyl amine hydrochloride,bis thiophene-2-methyl amine hydrochloride,bis thiophen-2-ylmethyl amine hydrochloride,di-2-thenylamine hydrochloride,1-thiophen-2-yl-n-thiophen-2-ylmethyl methanamine,hydrochloride,1-thiophen-2-yl-n-thiophen-2-yl methyl methanamine-hydrogen chloride 1/1
24212013
1-thiophen-2-yl-N-(thiophen-2-ylmethyl)methanamine;hydrochloride
C1=CSC(=C1)CNCC2=CC=CS2.Cl
336624-10-3
C10H12ClNS2
245.783
MFCD01812493
RTJOHEYKSNEPMI-UHFFFAOYSA-N
bis 2-thiophenemethyl amine hydrochloride,bis thiophene-2-methyl amine hydrochloride,bis thiophen-2-ylmethyl amine hydrochloride,di-2-thenylamine hydrochloride,1-thiophen-2-yl-n-thiophen-2-ylmethyl methanamine,hydrochloride,1-thiophen-2-yl-n-thiophen-2-yl methyl methanamine-hydrogen chloride 1/1
24212013
1-thiophen-2-yl-N-(thiophen-2-ylmethyl)methanamine;hydrochloride
C1=CSC(=C1)CNCC2=CC=CS2.Cl
Quantity 50g
Melting Point ~243°C (decomposition)
Formula Weight 245.8
Percent Purity 99%
Chemical Name or Material Bis(2-thiophenemethyl)amine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning