Alfa Aesar™ Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate, 95%

Manufacturer: Alfa Aesar™ H5337814

Quantity 25g

 View more versions of this product

Catalog No. AAH5337814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75203-51-9
    Molecular Formula C26H24O8Cl6
    Formula Weight 677.19
    MDL Number MFCD00012038
    Synonym Bis(2-carbopentyloxy-3,5,6-trichlorophenyl) oxalate
    Chemical Name or Material Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate

    Specifications

    Formula Weight 677.19
    Synonym Bis(2-carbopentyloxy-3,5,6-trichlorophenyl) oxalate
    Melting Point 76° to 80°C
    Quantity 25g
    Molecular Formula C26H24O8Cl6
    MDL Number MFCD00012038
    CAS 75203-51-9
    Chemical Name or Material Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit