Alfa Aesar™ Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine, 94%

Manufacturer: Alfa Aesar™ H2588206

Quantity 5g

 View more versions of this product

Catalog No. AAH2588206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 212713-08-1
    Molecular Formula C30H46ClO2P
    Formula Weight 505.13
    MDL Number MFCD08458881
    UN Number UN3261
    Chemical Name or Material Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine

    Specifications

    Formula Weight 505.13
    Molecular Formula C30H46ClO2P
    MDL Number MFCD08458881
    UN Number UN3261
    Health Hazard 1 Danger
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 212713-08-1
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine
    Assay 94%

    Description

    Applications
    It is used as a reactant for asymmetric hydrogenation with iridium catalysts, chemo selective reaction with deprotonated 2-hydoxy-3,3,N-trimethylbutanamide. It is also used as a reactant for synthesis of phosphine- containing amino acids and peptide-based catalyst ligands, nonracemic phosphine-containing amino acids for palladium- catalyzed asymmetric allylic substitution reactions.

    Solubility
    Reacts with water.

    Notes
    Air & moisture sensitive. Materials to avoid is oxidizing agents, bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit