Alfa Aesar™ Bis(3-aminopropyl)phenylphosphine

Manufacturer: Alfa Aesar™ 1042709

Quantity 10g

 View more versions of this product

Catalog No. AA1042709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6775-01-5
    Formula Weight 224.29
    MDL Number MFCD00014834
    UN Number UN3267
    Chemical Name or Material Bis(3-aminopropyl)phenylphosphine

    Specifications

    Formula Weight 224.29
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.5728
    Physical Form Liquid
    Quantity 10g
    Sensitivity Air Sensitive
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.
    UN Number UN3267
    Boiling Point 144°C (1mmHg)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Density 1.03g/mL
    MDL Number MFCD00014834
    CAS 6775-01-5
    Chemical Name or Material Bis(3-aminopropyl)phenylphosphine

    GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit