Alfa Aesar™ Bis(3-aminopropyl)phenylphosphine, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 4367509

Quantity 10g

 View more versions of this product

Catalog No. AA4367509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6775-01-5
    Molecular Formula (H2NC3H6)2P(C6H5)
    Formula Weight 224.29
    MDL Number MFCD00014834
    UN Number UN3267
    Chemical Name or Material Bis (3-aminopropyl)phenylphosphine

    Specifications

    Formula Weight 224.29
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Packaging ChemSeal™ bottles
    Refractive Index 1.5728
    Molecular Formula (H2NC3H6)2P(C6H5)
    MDL Number MFCD00014834
    Sensitivity Air Sensitive
    CAS 6775-01-5
    Boiling Point 144°C (1mmHg)
    Health Hazard 2 GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.
    Physical Form Liquid
    Density 1.03g/mL
    UN Number UN3267
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged under Argon
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Quantity 10g
    Chemical Name or Material Bis (3-aminopropyl)phenylphosphine

    GHS H Statement
    H314-H335-H319
    Causes severe skin burns and eye damage.
    May cause respiratory irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit