Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(3-aminopropyl)phenylphosphine, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 4367504

Quantity 2g

 View more versions of this product

Catalog No. AA4367504

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

6775-01-5
224.29
UN3267
(H2NC3H6)2P(C6H5)
MFCD00014834
Bis (3-aminopropyl)phenylphosphine

Specifications

6775-01-5
1.5728
1.03g/mL
224.29
UN3267
2g
Bis (3-aminopropyl)phenylphosphine
ChemSeal™ bottles
Liquid
(H2NC3H6)2P(C6H5)
MFCD00014834
144°C (1mmHg)
Air Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit