Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-bromophenyl)acetylene, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2789-89-1
C14H8Br2
336.02
MFCD00185147
FJQGIJIHOXZMMJ-UHFFFAOYSA-N
1,2-bis 4-bromophenyl ethyne,bis 4-bromophenyl acetylene,1-bromo-4-2-4-bromophenyl ethynyl benzene,bis-4-bromophenyl acetylene,1,2-bis 4-bromophenyl acetylene,benzene, 1,1'-1,2-ethynediyl bis 4-bromo,4,4'-dibromotolan,4,4'-dibromotolane,bis-p-bromophenyl acetylene,1 pound not2-bis 4-bromophenyl acetylene
3464242
1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
C1=CC(=CC=C1C#CC2=CC=C(C=C2)Br)Br
2789-89-1
C14H8Br2
336.02
MFCD00185147
FJQGIJIHOXZMMJ-UHFFFAOYSA-N
1,2-bis 4-bromophenyl ethyne,bis 4-bromophenyl acetylene,1-bromo-4-2-4-bromophenyl ethynyl benzene,bis-4-bromophenyl acetylene,1,2-bis 4-bromophenyl acetylene,benzene, 1,1'-1,2-ethynediyl bis 4-bromo,4,4'-dibromotolan,4,4'-dibromotolane,bis-p-bromophenyl acetylene,1 pound not2-bis 4-bromophenyl acetylene
3464242
1-bromo-4-[2-(4-bromophenyl)ethynyl]benzene
C1=CC(=CC=C1C#CC2=CC=C(C=C2)Br)Br
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 183°C to 187°C
Color White
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Linear Formula BrC6H4CCC6H4Br
Solubility Information Solubility in water: insoluble
Formula Weight 336.02
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Bis(4-bromophenyl)acetylene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning