Alfa Aesar™ Bis(4-bromophenyl) ether, 99%

Manufacturer: Alfa Aesar™ 1668406

Quantity 5g

 View more versions of this product

Catalog No. AA1668406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2050-47-7
    Formula Weight 328.01
    MDL Number MFCD00000095
    Chemical Name or Material Bis(4-bromophenyl) ether

    Specifications

    Formula Weight 328.01
    Melting Point 56° to 60°C
    RTECS Number KN0175000
    Physical Form Crystalline
    MDL Number MFCD00000095
    CAS 2050-47-7
    Color White
    Boiling Point 338° to 340°C
    TSCA TSCA
    Quantity 5g
    Chemical Name or Material Bis(4-bromophenyl) ether
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit