Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Bis(4-chlorophenylthio)methane, 99%
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 5g |
Melting Point | 45°C to 48°C |
Boiling Point | 193°C to 195°C (2mmHg) |
Beilstein | 2216758 |
Formula Weight | 301.26 |
Percent Purity | 99% |
Chemical Name or Material | Bis(4-chlorophenylthio)methane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures