Alfa Aesar™ Bis(4-fluorophenyl) disulfide, 98%

Manufacturer: Alfa Aesar™ H3258906

Quantity 5g

 View more versions of this product

Catalog No. AAH3258906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 405-31-2
    Molecular Formula C12H8F2S2
    Formula Weight 254.31
    MDL Number MFCD00816758
    Synonym 4,4'-Difluorodiphenyl disulfide
    Chemical Name or Material Bis(4-fluorophenyl) disulfide

    Specifications

    Formula Weight 254.31
    Molecular Formula C12H8F2S2
    MDL Number MFCD00816758
    CAS 405-31-2
    Synonym 4,4'-Difluorodiphenyl disulfide
    Health Hazard 1 Warning
    Boiling Point 141°C (1mmHg)
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Bis(4-fluorophenyl) disulfide
    Assay 98%

    Description

    Applications
    Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

    Solubility
    Soluble in methanol and acetone.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit