Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(4-fluorophenyl)methane, 98+%

Click to view available options
Quantity:
25g
100g
457-68-1
C13H10F2
204.22
MFCD00000360
DXQVFHQUHOFROC-UHFFFAOYSA-N
bis 4-fluorophenyl methane,4,4'-difluorodiphenylmethane,1,1'-methylenebis 4-fluorobenzene,1-fluoro-4-4-fluorophenyl methyl benzene,benzene, 1,1'-methylenebis 4-fluoro,4-fluoro-1-4-fluorophenyl methyl benzene,4,4'-difluorodiphenylamine,bis-4-fluorophenyl methane,4,4'-difluorodiphenyl methane,1,1-methylenebis 4-fluorobenzene
96094
1-fluoro-4-[(4-fluorophenyl)methyl]benzene
C1=CC(=CC=C1CC2=CC=C(C=C2)F)F
457-68-1
C13H10F2
204.22
MFCD00000360
DXQVFHQUHOFROC-UHFFFAOYSA-N
bis 4-fluorophenyl methane,4,4'-difluorodiphenylmethane,1,1'-methylenebis 4-fluorobenzene,1-fluoro-4-4-fluorophenyl methyl benzene,benzene, 1,1'-methylenebis 4-fluoro,4-fluoro-1-4-fluorophenyl methyl benzene,4,4'-difluorodiphenylamine,bis-4-fluorophenyl methane,4,4'-difluorodiphenyl methane,1,1-methylenebis 4-fluorobenzene
96094
1-fluoro-4-[(4-fluorophenyl)methyl]benzene
C1=CC(=CC=C1CC2=CC=C(C=C2)F)F
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Boiling Point 265°C
Flash Point 134°C (273°F)
Melting Point 27°C to 30°C
Quantity 100g
Beilstein 1956387
Formula Weight 204.22
Percent Purity ≥98%
Chemical Name or Material Bis(4-fluorophenyl)methane