Alfa Aesar™ Bis(4-methoxyphenyl)phenylphosphine, 95%

Manufacturer: Alfa Aesar™ B2535614

Quantity 25g

 View more versions of this product

Catalog No. AAB2535614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14180-51-9
    Molecular Formula C20H19O2P
    Formula Weight 322.34
    MDL Number MFCD00048993
    Chemical Name or Material Bis(4-methoxyphenyl)phenylphosphine

    Specifications

    Formula Weight 322.34
    Molecular Formula C20H19O2P
    MDL Number MFCD00048993
    Melting Point 88° to 90°C
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 14180-51-9
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material Bis(4-methoxyphenyl)phenylphosphine
    Assay 95%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit