Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-methyl-1-piperazinylthiocarbonyl) disulfide, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
20231-01-0
C12H22N4S4
350.576
MFCD01729803
OOBDBFPYJXJEHC-UHFFFAOYSA-N
4-methylpiperazino-thiuramdisulfide,bis 4-methyl-1-piperazinyl thiocarbonyl disulfide,bis 4-methyl-1-piperazinylthiocarbonyl disulfide,piperazine, 1,1'-dithiodicarbonothioyl bis 4-methyl,disulfide, bis 4-methyl-1-piperazinyl thiocarbonyl,5-23-01-00114 beilstein handbook reference,bis 4-methylpiperazine-1-thiocarbonyl disulfane,bis 4-methylpiperazino thiocarbonyl persulfide,4-methylpiperazine-1-carbothioic dithioperoxyanhydride
30054
(4-methylpiperazine-1-carbothioyl)sulfanyl 4-methylpiperazine-1-carbodithioate
CN1CCN(CC1)C(=S)SSC(=S)N2CCN(CC2)C
20231-01-0
C12H22N4S4
350.576
MFCD01729803
OOBDBFPYJXJEHC-UHFFFAOYSA-N
4-methylpiperazino-thiuramdisulfide,bis 4-methyl-1-piperazinyl thiocarbonyl disulfide,bis 4-methyl-1-piperazinylthiocarbonyl disulfide,piperazine, 1,1'-dithiodicarbonothioyl bis 4-methyl,disulfide, bis 4-methyl-1-piperazinyl thiocarbonyl,5-23-01-00114 beilstein handbook reference,bis 4-methylpiperazine-1-thiocarbonyl disulfane,bis 4-methylpiperazino thiocarbonyl persulfide,4-methylpiperazine-1-carbothioic dithioperoxyanhydride
30054
(4-methylpiperazine-1-carbothioyl)sulfanyl 4-methylpiperazine-1-carbodithioate
CN1CCN(CC1)C(=S)SSC(=S)N2CCN(CC2)C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 250mg
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 350.59
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Bis(4-methyl-1-piperazinylthiocarbonyl) disulfide, 98%
Show More Show Less