Alfa Aesar™ Bis(4-morpholinyl)methane, 98%

Manufacturer: Alfa Aesar™ H5113314

Quantity 25g

 View more versions of this product

Catalog No. AAH5113314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5625-90-1
    Molecular Formula C9H18N2O2
    Formula Weight 186.25
    MDL Number MFCD00023369
    Synonym 4,4'-Methylenebismorpholine; 4,4'-Dimorpholinylmethane
    Chemical Name or Material Bis(4-morpholinyl)methane

    Specifications

    Formula Weight 186.25
    Molecular Formula C9H18N2O2
    MDL Number MFCD00023369
    CAS 5625-90-1
    Synonym 4,4'-Methylenebismorpholine; 4,4'-Dimorpholinylmethane
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H318-H315
    Causes serious eye damage.
    Causes skin irritation.
    Boiling Point 122° to 124°C (12mmHg)
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280b-P305+P351+P338-P337+P313
    Wear protective gloves and eye protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material Bis(4-morpholinyl)methane
    Assay 98%

    Description

    Applications
    It is employed as intermediate for pharmaceutical.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H318-H315
    Causes serious eye damage.
    Causes skin irritation.

    GHS P Statement
    P280b-P305+P351+P338-P337+P313
    Wear protective gloves and eye protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit