Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-nitrophenyl) carbonate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5070-13-3
C13H8N2O7
304.21
MFCD00007322
ACBQROXDOHKANW-UHFFFAOYSA-N
bis 4-nitrophenyl carbonate,bis p-nitrophenyl carbonate,4,4'-dinitrodiphenyl carbonate,carbonic acid, bis 4-nitrophenyl ester,p,p'-dinitrodiphenylcarbonate,carbonic acid bis 4-nitrophenyl ester,carbonic acid, bis p-nitrophenyl ester,npc bis 4-nitrophenyl carbonate
78756
bis(4-nitrophenyl) carbonate
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
5070-13-3
C13H8N2O7
304.21
MFCD00007322
ACBQROXDOHKANW-UHFFFAOYSA-N
bis 4-nitrophenyl carbonate,bis p-nitrophenyl carbonate,4,4'-dinitrodiphenyl carbonate,carbonic acid, bis 4-nitrophenyl ester,p,p'-dinitrodiphenylcarbonate,carbonic acid bis 4-nitrophenyl ester,carbonic acid, bis p-nitrophenyl ester,npc bis 4-nitrophenyl carbonate
78756
bis(4-nitrophenyl) carbonate
C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]
Coupling reagent
Melting Point 139°C to 141°C
Color Brown to Green or Yellow
NMR Consistent with structure
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula (O2NC6H4O)2CO
Beilstein 06, I, 120
Fieser 01,330
Formula Weight 304.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Bis(4-nitrophenyl) carbonate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-775-4