Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, ≥99% (metals basis)

Manufacturer:  Alfa Aesar™ H55188MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55188MD


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

131717-99-2
MFCD02683472
Electrophoresis Grade
564.52
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate

Specifications

131717-99-2
564.52
White
250mg
Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate
Electrophoresis Grade
Powder
MFCD02683472
237° to 240°C
Light Sensitive
≥99%, (metals basis)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit