Alfa Aesar™ Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate, Electronic grade, ≥99% (metals basis)

Manufacturer: Alfa Aesar™ H5597603

Quantity 1g

 View more versions of this product

Catalog No. AAH5597603

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 84563-54-2
    Formula Weight 542.4
    MDL Number MFCD02683471
    Synonym Bis(4-tert-butylphenyl)iodonium triflate
    Chemical Name or Material Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate
    Grade Electrophoresis Grade

    Specifications

    Formula Weight 542.4
    Melting Point 162° to 166°C
    Physical Form Powder
    MDL Number MFCD02683471
    CAS 84563-54-2
    Color White
    Synonym Bis(4-tert-butylphenyl)iodonium triflate
    Quantity 1g
    Sensitivity Light Sensitive
    Chemical Name or Material Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate
    Assay ≥99%, (metals basis)
    Grade Electrophoresis Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit