Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate, ≥97%

Click to view available options
Quantity:
250mg
1g
32679-03-1
32679-03-1
Quantity 1g
Sensitivity Air Sensitive
Color Yellow
Formula Weight 469.31
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material Bis(acetonitrile)(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning