Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, ≥97%

Click to view available options
Quantity:
1g
5g
32679-02-0
C12H18BF4N2Rh-
379.999
MFCD06798085
ZIBLHOBPBGAKNV-SUESZSCISA-N
bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate,rh cod mecn 2 bf4,bis acetonitrile 1,5-cyclooctadiene rhodium i te,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate
11132655
acetonitrile;(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC#N.CC#N.C1CC=CCCC=C1.[Rh]
32679-02-0
C12H18BF4N2Rh-
379.999
MFCD06798085
ZIBLHOBPBGAKNV-SUESZSCISA-N
bis acetonitrile 1,5-cyclooctadiene rhodium 1 tetrafluoroborate,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate,rh cod mecn 2 bf4,bis acetonitrile 1,5-cyclooctadiene rhodium i te,bis acetonitrile 1,5-cyclooctadiene rhodium i tetrafluoroborate
11132655
acetonitrile;(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[B-](F)(F)(F)F.CC#N.CC#N.C1CC=CCCC=C1.[Rh]
Sensitivity Air Sensitive
Color Yellow
Quantity 1g
Formula Weight 379.99
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material Bis(acetonitrile)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning