Alfa Aesar™ Bis(acetonitrile)dichloropalladium(II), Pd 40.5%

Olefin isomerization, preparation of acetals and hemiacetal esters

Manufacturer: Alfa Aesar™ 1000206

Quantity 5g

 View more versions of this product

Catalog No. AA1000206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14592-56-4
    Formula Weight 259.41
    MDL Number MFCD00013122
    UN Number UN3439
    Synonym cis-Dichlorobis(acetonitrile)palladium(II)
    Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)

    Specifications

    Formula Weight 259.41
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    MDL Number MFCD00013122
    CAS 14592-56-4
    UN Number UN3439
    Health Hazard 1 Danger
    Synonym cis-Dichlorobis(acetonitrile)palladium(II)
    Quantity 5g
    Solubility Information Soluble in acetonitrile, acetone, chloroform
    Concentration Pd 40.5%
    Odor Odorless
    Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit