Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bis(acetonitrile)dichloropalladium(II), Pd 40.5%

Click to view available options
Quantity:
1g
5g
14592-56-4
MFCD00013122
cis-Dichlorobis(acetonitrile)palladium(II)
14592-56-4
MFCD00013122
cis-Dichlorobis(acetonitrile)palladium(II)

Olefin isomerization, preparation of acetals and hemiacetal esters

Odor Odorless
Quantity 1g
UN Number UN3439
Concentration or Composition (by Analyte or Components) Pd 40.5%
Solubility Information Soluble in acetonitrile, acetone, chloroform
Formula Weight 259.41
Physical Form Powder
Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P362-P501c

H301+H311+H331-H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 238-637-3

TSCA : No

Recommended Storage : Ambient temperatures