Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(acetonitrile)palladium(II) chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
500mg
5g
Packaging:
Glass bottle
14592-56-4
C4H6Cl2N2Pd
259.4
MFCD00013122
14592-56-4
C4H6Cl2N2Pd
259.4
MFCD00013122
Infrared Spectrum Authentic
Melting Point 300°C
Packaging Glass bottle
Quantity 500mg
Color Brown-Rust to Orange
Elemental Analysis 98.5% min.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3CN)2PdCl2
Fieser 13,33; 15,28; 16,25; 17,30; 01,1015; 03,254; 04,435
Solubility Information Solubility in water: insoluble
Formula Weight 259.4
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material Bis(acetonitrile)palladium(II) chloride, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-637-3