Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Bis-acrylamide Solution (2% w/v/Electrophoresis), Fisher BioReagents
Click to view available options
Quantity:
250 mL
Packaging:
Amber Glass
Description
Bis-Acrylamide Solution is a cross-linker commonly used in the preparation of polyacrylamide electrophoresis gels.
Specifications
Physical Form | Liquid |
Color | Colorless |
Quantity | 250 mL |
Formula Weight | 154.08 |
Packaging | Amber Glass |
Grade | Electrophoresis |
Chemical Name or Material | Bis-Acrylamide Solution, 2% w/v |
Safety and Handling

Recommended Storage : 4°C